Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133924.3 | NP_598685.2 | 3 | 1107 | ||
GenBank mRNA | AK141663.1 | - | 3 | 1108 | ||
AK161207.1 | - | 1 | 706 | |||
AK168473.1 | - | 3 | 1096 | |||
BC017623.1 | - | 1 | 154 | |||
BC037155.1 | - | 1 | 43 | |||
BC094371.1 | - | 2 | 1803 | |||
BC117542.1 | - | 4 | 1327 | 40105600 |