Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133932.2 | NP_598693.1 | 4 | 770 | ||
GenBank mRNA | AF383154.1 | - | 4 | 589 | ||
AK078906.1 | - | 4 | 1090 | |||
AK211996.1 | - | 1 | 99 | |||
BC023879.1 | - | 4 | 957 | |||
BC026630.1 | - | 4 | 770 | 4983657 |