Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207210.2 | NP_997093.2 | 3 | 235 | ||
GenBank mRNA | AK077117.1 | - | 3 | 236 | ||
BC052324.1 | - | 2 | 289 | 6315342 |