Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175315.1 | NP_780524.1 | 2 | 457 | ||
GenBank mRNA | AK049385.1 | - | 2 | 372 | ||
BC099491.1 | - | 2 | 363 | |||
BC125404.1 | - | 2 | 457 | 40129919 | ||
BC125406.1 | - | 2 | 457 | 40129921 |