Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014981.1 | NP_001014981.1 | 12, 13 | 1827 | ||
GenBank mRNA | AF188123.1 | - | 11, 12 | 1589 | ||
AF188124.1 | - | 11, 12 | 1589 | |||
AK122304.1 | - | 4, 5 | 716 | |||
AK154451.1 | - | 12, 13 | 1807 | |||
BC026618.1 | - | 12, 13 | 1746 | |||
BC125573.1 | - | 12, 13 | 1695 | 40130088 | ||
BC137785.1 | - | 12, 13 | 1695 | 8860804 |