Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134034.2 | NP_598795.2 | 2 | 554 | ||
GenBank mRNA | AK037253.1 | - | 2 | 486 | ||
AK041513.1 | - | 2 | 499 | |||
AK044742.1 | - | 2 | 495 | |||
AK090014.1 | - | 2 | 530 | |||
AK129344.1 | - | 2 | 465 | |||
AK132660.1 | - | 2 | 76 | |||
AK165940.1 | - | 2 | 478 | |||
AK166627.1 | - | 2 | 480 | |||
BC006870.1 | - | 2 | 465 | 3595439 | ||
CD350450.1 | - | 2 | 554 |