Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036623.1 | - | 20 | 2521 | ||
AK151178.1 | - | 20 | 2455 | |||
AK153039.1 | - | 20 | 2455 | |||
AK169948.1 | - | 20 | 2752 | |||
AY753194.1 | - | 18 | 2211 | |||
BC028998.1 | - | 20 | 2451 | 5356085 | ||
DQ295832.1 | - | 18 | 2223 |