Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033213.1 | NP_001028385.1 | 6 | 879 | ||
GenBank mRNA | AK046740.1 | - | 6 | 848 | ||
AK148363.1 | - | 6 | 851 |