Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam208b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134063.3 | NP_598824.3 | 17, 18 | 7416 | ||
GenBank mRNA | AK128934.1 | - | 4, 5 | 2733 | ||
AK131707.1 | - | 6, 7 | 4812 | |||
AK220437.1 | - | 17, 18 | 7404 | |||
BC016423.1 | - | 4, 5 | 2100 | |||
BC045617.1 | - | 4, 5 | 2439 | |||
BC085160.1 | - | 8, 9 | 6159 |
Gene Symbol: Gdi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK163431.1 | - | 1719 |