Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053180.2 | NP_444410.1 | 5 | 591 | ||
GenBank mRNA | AY005133.1 | - | 5 | 602 | ||
BC031907.1 | - | 5 | 591 | 5362198 | ||
BC058353.1 | - | 872 |