Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cggbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178647.2 | NP_848762.1 | 3 | 491 | ||
GenBank mRNA | AK049631.1 | - | 3 | 569 | ||
AK052195.1 | - | 3 | 559 | |||
AK083450.1 | - | 3 | 496 | |||
AK086053.1 | - | 3 | 570 | |||
AK194750.1 | - | 1 | 31 | |||
BC043097.1 | - | 3 | 491 | 6405954 | ||
BC057053.1 | - | 3 | 545 | 6827183 |