Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025572.1 | NP_001020743.1 | 9 | 1399 | ||
GenBank mRNA | AK134530.1 | - | 4 | 578 | ||
AK153850.1 | - | 8 | 1308 | |||
BC023046.1 | - | 9 | 1270 | |||
BC064777.1 | - | 8 | 1314 | |||
BC080825.1 | - | 8 | 1322 |