Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166474.1 | NP_001159946.1 | 2 | 178 | ||
NM_178926.3 | NP_849257.1 | 2 | 364 | |||
GenBank mRNA | AK078549.1 | - | 2 | 295 | ||
AK089265.1 | - | 2 | 422 | |||
AK145854.1 | - | 2 | 217 | |||
AK170944.1 | - | 2 | 310 | |||
BC026472.1 | - | 2 | 364 | 4487505 | ||
BC039944.1 | - | 2 | 142 | |||
BC058424.1 | - | 2 | 159 |