Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148944.4 | NP_683746.1 | 4 | 376 | ||
GenBank mRNA | AF492840.1 | - | 4 | 361 | ||
AK140071.1 | - | 4 | 378 | |||
AY574269.1 | - | 4 | 327 | |||
BC094889.1 | - | 4 | 422 | |||
BC138851.1 | - | 4 | 375 | 8861873 |