Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024241.2 | NP_077203.2 | 2 | 503 | ||
GenBank mRNA | AK016916.1 | - | 4 | 704 | ||
AK050683.1 | - | 2 | 489 | |||
AK084967.1 | - | 2 | 504 | |||
AK135307.1 | - | 2 | 504 | |||
BB659870.1 | - | 2 | 504 | |||
BC067395.1 | - | 2 | 495 | 30460089 |