Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026815.2 | NP_081091.1 | 6 | 598 | ||
GenBank mRNA | AF262335.1 | - | 5 | 570 | ||
AK004014.1 | - | 6 | 599 | |||
BC132047.1 | - | 5 | 590 | 40130324 | ||
BC138390.1 | - | 5 | 590 | 8861410 |