Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289730.1 | NP_001276659.1 | 10 | 1482 | ||
NM_001289731.1 | NP_001276660.1 | 13 | 2038 | |||
NM_133197.4 | NP_573460.2 | 10 | 1482 | |||
GenBank mRNA | AB052945.1 | - | 10 | 1425 | ||
AB101616.1 | - | 10 | 1425 | |||
AB244989.1 | - | 1 | 36 | |||
AK032122.1 | - | 10 | 1483 | |||
AK045424.1 | - | 13 | 2040 | |||
BC125435.1 | - | 10 | 1384 | 40129950 |