Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145615.4 | NP_663590.3 | 6 | 651 | ||
GenBank mRNA | AK028118.1 | - | 6 | 652 | ||
AK032830.1 | - | 6 | 604 | |||
AK133525.1 | - | 6 | 597 | |||
AK167572.1 | - | 6 | 624 | |||
AK168321.1 | - | 6 | 612 | |||
AK218235.1 | - | 5 | 342 | |||
BC003432.1 | - | 6 | 542 | 2646522 | ||
BC096645.1 | - | 6 | 510 | 1245213 |