Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133199.2 | NP_573462.2 | 1 | 131 | ||
GenBank mRNA | AJ278787.1 | - | 1 | 131 | ||
BC129805.1 | - | 1 | 129 |