Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033617.3 | NP_291095.1 | 1, 2 | 155 | ||
GenBank mRNA | AB060654.1 | - | 1, 2 | 116 | ||
AF356006.1 | - | 1, 2 | 159 | |||
AK009495.1 | - | 1, 2 | 147 | |||
AK021272.1 | - | 1, 2 | 135 | |||
AK151365.1 | - | 1, 2 | 127 | |||
AK151847.1 | - | 1, 2 | 143 | |||
AK152330.1 | - | 1, 2 | 160 | |||
AK152823.1 | - | 1, 2 | 143 | |||
AK200953.1 | - | 1, 2 | 69 | |||
AK217120.1 | - | 1, 2 | 111 | |||
BC030393.1 | - | 1, 2 | 140 | 4922226 |