Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054060.1 | NP_473401.1 | 2 | 386 | ||
GenBank mRNA | BC100352.1 | - | 2 | 386 | 30715445 |