Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007422.3 | NP_031448.2 | 6 | 620 | ||
GenBank mRNA | AK004877.1 | - | 6 | 573 | ||
AK010263.1 | - | 6 | 595 | |||
AK028060.1 | - | 6 | 620 | |||
AK043273.1 | - | 6 | 613 | |||
AK148420.1 | - | 6 | 602 | |||
AK170279.1 | - | 6 | 606 | |||
BC139404.1 | - | 6 | 532 | 8862426 | ||
BC139417.1 | - | 6 | 532 | 8862439 |