Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009658.3 | NP_033788.3 | 2 | 228 | ||
GenBank mRNA | AK028079.1 | - | 2 | 183 | ||
AK139250.1 | - | 2 | 183 | |||
AK145392.1 | - | 2 | 184 | |||
AK149861.1 | - | 2 | 218 | |||
AK150197.1 | - | 2 | 183 | |||
AK153949.1 | - | 2 | 212 | |||
AK164682.1 | - | 2 | 183 | |||
AK170663.1 | - | 2 | 230 | |||
BC004725.1 | - | 2 | 175 | 3582788 | ||
BC021655.1 | - | 2 | 221 | 3708927 | ||
BC085310.1 | - | 2 | 160 | 6485416 | ||
BY749711.1 | - | 2 | 228 | |||
L39795.1 | - | 2 | 160 | |||
U29152.1 | - | 2 | 179 |
Gene Symbol: Csnk1g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK195453.1 | - | 24 |