Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cts3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026906.3 | NP_081182.2 | 6 | 870 | ||
NM_054092.2 | NP_473433.2 | 6 | 795 | |||
GenBank mRNA | AK005389.1 | - | 6 | 793 | ||
AY034574.1 | - | 6 | 809 | |||
BC099388.1 | - | 6 | 764 | 30786828 |