Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110505.1 | NP_001103975.1 | 10, 11 | 1446 | ||
NM_007446.2 | NP_031472.2 | 10, 11 | 1555 | |||
GenBank mRNA | AA616210.1 | - | 3, 4 | 338 | ||
AK029642.1 | - | 10, 11 | 1478 | |||
BB462169.1 | - | 1, 2 | 95 | |||
BC004698.1 | - | 6, 7 | 789 | |||
BC009121.1 | - | 10, 11 | 1429 | 3710941 | ||
J00356.1 | - | 9, 10 | 1399 | |||
V00717.1 | - | 10, 11 | 1444 | |||
V00719.1 | - | 10, 11 | 1555 | |||
V00720.1 | - | 10, 11 | 1446 |
Gene Symbol: Amy2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M11895.1 | - | 181 |
Gene Symbol: Amy2a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X02577.1 | - | 1358 |