Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109045.1 | NP_001102515.1 | 3 | 468 | ||
NM_007474.2 | NP_031500.1 | 3 | 471 | |||
GenBank mRNA | AF018952.1 | - | 3 | 453 | ||
AK146660.1 | - | 2 | 150 | |||
BC010982.1 | - | 3 | 413 | 4224850 | ||
BY713653.1 | - | 3 | 473 | |||
CF617211.1 | - | 3 | 374 |