Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109045.1 | NP_001102515.1 | 4 | 691 | ||
NM_007474.2 | NP_031500.1 | 4 | 694 | |||
GenBank mRNA | AF018952.1 | - | 4 | 676 | ||
AK146660.1 | - | 3 | 373 | |||
BC010982.1 | - | 4 | 636 | 4224850 | ||
CF617211.1 | - | 4 | 597 |