Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arntl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243048.1 | NP_001229977.1 | 8 | 809 | ||
NM_007489.4 | NP_031515.1 | 11 | 1102 | |||
GenBank mRNA | AB012601.1 | - | 8 | 603 | ||
AB012602.1 | - | 7 | 554 | |||
AB014494.1 | - | 11 | 988 | |||
AB015203.1 | - | 7 | 582 | |||
AK143267.1 | - | 11 | 1080 | |||
AK146628.1 | - | 11 | 1102 | |||
AK147145.1 | - | 11 | 1102 | |||
AY690602.1 | - | 8 | 575 | |||
BC011080.1 | - | 8 | 760 | 3661605 | ||
BC025973.1 | - | 9 | 853 | 3980719 |