Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007503.3 | NP_031529.2 | 3 | 185 | ||
NM_052823.2 | NP_439888.1 | 3 | 258 | |||
GenBank mRNA | AA396010.1 | - | 3 | 206 | ||
AI316264.1 | - | 3 | 169 | |||
AK002215.1 | - | 3 | 173 | |||
AK002228.1 | - | 3 | 186 | |||
AK007105.1 | - | 1 | 60 | |||
AY626243.1 | - | 3 | 307 | |||
AY626244.1 | - | 3 | 258 | |||
AY626245.1 | - | 3 | 343 | |||
BC024614.1 | - | 3 | 174 | 4222609 | ||
BC069892.1 | - | 3 | 182 | |||
BU756853.1 | - | 4 | 368 | |||
CA491597.1 | - | 3 | 424 | |||
CB600439.1 | - | 3 | 218 | |||
CB600958.1 | - | 3 | 393 | |||
X70060.1 | - | 2 | 96 |