Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077265.2 | NP_001070733.1 | 6 | 1094 | ||
NM_001077266.2 | NP_001070734.1 | 5 | 1037 | |||
NM_001077267.2 | NP_001070735.1 | 5 | 1037 | |||
NM_007516.3 | NP_031542.2 | 6 | 1094 | |||
GenBank mRNA | AK077409.1 | - | 6 | 1082 | ||
BC011172.1 | - | 6 | 769 | |||
BC049098.1 | - | 6 | 1094 | 5717286 | ||
CA451481.1 | - | 4 | 375 | |||
CF751545.1 | - | 3 | 245 | |||
CX567003.1 | - | 2 | 69 |