Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285453.1 | NP_001272382.1 | 2, 3 | 234 | ||
NM_007522.3 | NP_031548.1 | 2, 3 | 751 | |||
GenBank mRNA | AK131666.1 | - | 2, 3 | 196 | ||
AK151792.1 | - | 2, 3 | 199 | |||
AK169002.1 | - | 2, 3 | 196 | |||
BC006762.1 | - | 2, 3 | 719 | 3257589 | ||
L37296.1 | - | 2, 3 | 759 |