Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd24a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009846.2 | NP_033976.1 | 2 | 192 | ||
GenBank mRNA | AK133869.1 | - | 2 | 191 | ||
AK168111.1 | - | 2 | 191 | |||
AK169613.1 | - | 2 | 189 | |||
BC075622.1 | - | 2 | 172 | 6415159 | ||
M58661.1 | - | 2 | 179 | |||
X53825.1 | - | 2 | 130 |