Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009930.2 | NP_034060.2 | 1, 2 | 293 | ||
GenBank mRNA | AK013329.1 | - | 1, 2 | 201 | ||
AK029212.1 | - | 1, 2 | 197 | |||
AK031544.1 | - | 1, 2 | 166 | |||
AK041115.1 | - | 1, 2 | 201 | |||
AK048546.1 | - | 1, 2 | 201 | |||
AK147542.1 | - | 1, 2 | 199 | |||
AK157762.1 | - | 1, 2 | 201 | |||
AK159763.1 | - | 1, 2 | 201 | |||
AK159920.1 | - | 1, 2 | 201 | |||
AK159985.1 | - | 1, 2 | 201 | |||
AK160048.1 | - | 1, 2 | 201 | |||
AK160107.1 | - | 1, 2 | 200 | |||
AK160818.1 | - | 1, 2 | 201 | |||
AK220558.1 | - | 1, 2 | 216 | |||
BC043089.1 | - | 1, 2 | 168 | 6418299 | ||
BC052398.1 | - | 1, 2 | 185 | 5720872 | ||
BC058724.1 | - | 1, 2 | 202 | 5705762 | ||
BY139268.1 | - | 1, 2 | 293 | |||
M18933.1 | - | 1, 2 | 102 |