Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007737.2 | NP_031763.2 | 52 | 4323 | ||
GenBank mRNA | AK084285.1 | - | 14 | 1140 | ||
AK084538.1 | - | 11 | 1107 | |||
AK084803.1 | - | 52 | 3962 | |||
AK132413.1 | - | 52 | 4105 | |||
AK139130.1 | - | 52 | 3898 | |||
AK147220.1 | - | 52 | 4105 | |||
AK147328.1 | - | 52 | 4324 | |||
AK151929.1 | - | 52 | 4107 | |||
AK160008.1 | - | 52 | 3897 | |||
BC043696.1 | - | 26 | 2138 | |||
BC055077.1 | - | 52 | 4074 | 30059991 | ||
L02918.1 | - | 52 | 3955 |