Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277219.1 | NP_001264148.1 | 2 | 436 | ||
NM_133656.5 | NP_598417.2 | 2 | 436 | |||
GenBank mRNA | AF239673.1 | - | 2 | 306 | ||
AK028179.1 | - | 2 | 413 | |||
AK028488.1 | - | 2 | 386 | |||
AK036905.1 | - | 2 | 387 | |||
AK052795.1 | - | 2 | 395 | |||
AK076097.1 | - | 2 | 131 | |||
AK088519.1 | - | 2 | 407 | |||
AK139089.1 | - | 2 | 392 | |||
AK152549.1 | - | 2 | 410 | |||
AK154987.1 | - | 2 | 421 | |||
AK161356.1 | - | 2 | 386 | |||
AK161761.1 | - | 2 | 378 | |||
BC012216.1 | - | 2 | 372 | 3992341 | ||
BC031149.1 | - | 2 | 369 | 4975654 | ||
S72408.1 | - | 2 | 361 |