Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021541.3 | NP_067516.1 | 3 | 263 | ||
GenBank mRNA | AJ272227.1 | - | 3 | 255 | ||
AK021374.1 | - | 3 | 237 | |||
AY160973.1 | - | 3 | 228 | |||
BC052746.1 | - | 3 | 226 | 6824801 | ||
BC056989.1 | - | 3 | 217 | 6824245 | ||
CF740624.1 | - | 3 | 262 |