Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010062.3 | NP_034192.1 | 1, 2 | 244 | ||
GenBank mRNA | AK145192.1 | - | 1, 2 | 245 | ||
BC058609.1 | - | 1, 2 | 191 | 5397016 | ||
BQ829838.1 | - | 1, 2 | 230 |