Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146346.1 | NP_001139818.1 | 4 | 554 | ||
NM_010129.2 | NP_034259.2 | 4 | 525 | |||
GenBank mRNA | AF011750.1 | - | 4 | 386 | ||
AK146939.1 | - | 4 | 557 | |||
BC001999.1 | - | 4 | 370 | 3485948 | ||
CK022835.1 | - | 4 | 524 | |||
U87948.1 | - | 4 | 503 |