Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252662.1 | NP_001239591.1 | 10 | 1226 | ||
NM_001252663.1 | NP_001239592.1 | 10 | 1226 | |||
NM_001252664.1 | NP_001239593.1 | 9 | 1053 | |||
NM_001252665.1 | NP_001239594.1 | 9 | 952 | |||
NM_001252666.1 | NP_001239595.1 | 8 | 985 | |||
NM_013514.4 | NP_038542.1 | 11 | 1167 | |||
GenBank mRNA | AF079846.3 | - | 9 | 896 | ||
AF155547.1 | - | 9 | 896 | |||
AK143840.1 | - | 10 | 1220 | |||
AK149301.1 | - | 9 | 863 | |||
AK155927.1 | - | 10 | 1221 | |||
AK158744.1 | - | 9 | 984 | |||
AK162866.1 | - | 9 | 904 | |||
AK165204.1 | - | 1219 | ||||
AK165294.1 | - | 10 | 1218 | |||
BC016897.1 | - | 11 | 1167 | 4240747 | ||
BC037021.1 | - | 8 | 972 | 5366040 |