Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007957.2 | NP_031983.2 | 2 | 399 | ||
GenBank mRNA | AF004211.1 | - | 1 | 487 | ||
AF017734.1 | - | 1 | 116 | |||
AF017735.1 | - | 2 | 199 | |||
AF085715.1 | - | 2 | 400 | |||
DQ059551.1 | - | 1 | 436 |