Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080465.2 | NP_536713.1 | 3 | 777 | ||
GenBank mRNA | AF357240.1 | - | 3 | 777 | ||
AF533008.1 | - | 3 | 777 | |||
AK033158.1 | - | 6 | 2281 | |||
AK039033.1 | - | 3 | 1463 | |||
AK050390.1 | - | 3 | 1033 | |||
AK087908.1 | - | 3 | 1122 | |||
AK149082.1 | - | 3 | 1465 | |||
AY123778.1 | - | 3 | 852 | |||
AY280964.1 | - | 3 | 777 | |||
BC125475.1 | - | 3 | 803 | 40129990 | ||
BC137656.1 | - | 3 | 803 | 8860675 |