Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161373.1 | NP_001154845.1 | 9 | 1813 | ||
NM_001161374.1 | NP_001154846.1 | 9 | 1813 | |||
NM_007977.2 | NP_032003.2 | 9 | 1813 | |||
GenBank mRNA | BC138347.1 | - | 9 | 1694 | 8861367 | |
BC145245.1 | - | 9 | 1694 | 9053797 | ||
L05573.1 | - | 9 | 1811 |