Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fpgs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010236.2 | NP_034366.2 | 15 | 1584 | ||
GenBank mRNA | AK041952.1 | - | 13 | 1726 | ||
AK157040.1 | - | 15 | 1545 | |||
AK181115.1 | - | 2 | 213 | |||
BC005484.1 | - | 15 | 1527 | 3485401 | ||
BM944126.1 | - | 6 | 583 | |||
U32197.1 | - | 15 | 1541 | |||
U33557.1 | - | 15 | 1561 |