Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019521.2 | NP_062394.2 | 6 | 711 | ||
GenBank mRNA | AK086187.1 | - | 6 | 705 | ||
BC005444.1 | - | 6 | 666 | 3592398 | ||
X59846.1 | - | 6 | 682 |