Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008138.4 | NP_032164.2 | 4 | 499 | ||
GenBank mRNA | AK157998.1 | - | 4 | 503 | ||
AK159222.1 | - | 4 | 503 | |||
AK160186.1 | - | 4 | 495 | |||
AK167388.1 | - | 4 | 501 | |||
AK185434.1 | - | 1 | 73 | |||
BC065159.1 | - | 4 | 486 | 5717305 | ||
M13963.1 | - | 4 | 471 | |||
S79607.1 | - | 3 | 256 |