Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfdn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185182.1 | NP_001172111.1 | 5 | 368 | ||
NM_010385.2 | NP_034515.1 | 4 | 572 | |||
GenBank mRNA | AA023721.1 | - | 4 | 348 | ||
AK010679.1 | - | 4 | 562 | |||
AK076015.1 | - | 4 | 555 | |||
AV446044.1 | - | 3 | 227 |