Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010452.3 | NP_034582.1 | 5 | 1091 | ||
GenBank mRNA | AK053218.1 | - | 1 | 232 | ||
AK076761.1 | - | 1 | 145 | |||
BC096612.1 | - | 4 | 1067 | 6476230 | ||
U56399.1 | - | 2 | 600 | |||
Y11717.1 | - | 5 | 1141 |