Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008266.5 | NP_032292.3 | 1 | 82 | ||
GenBank mRNA | AK017686.1 | - | 1 | 83 | ||
BC103597.1 | - | 1 | 79 | 40046066 | ||
BC103598.1 | - | 1 | 79 | 40046076 | ||
BC103605.1 | - | 1 | 79 | 40046069 | ||
BC103606.1 | - | 1 | 79 | 40046068 |