Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010460.2 | NP_034590.2 | 1 | 95 | ||
GenBank mRNA | AK136706.1 | - | 1 | 95 | ||
X06762.1 | - | 2177 |