Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008377.2 | NP_032403.2 | 19 | 3588 | ||
GenBank mRNA | AK133771.1 | - | 6 | 1156 | ||
BC141300.1 | - | 19 | 3380 | 9055912 | ||
BC145623.1 | - | 18 | 3233 | 9054191 | ||
CN699905.1 | - | 3 | 463 | |||
D78572.1 | - | 19 | 3581 |